Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAR1/Thrombin Receptor Antibody (731115), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348115 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348115 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348115 Supplier Novus Biologicals Supplier No. FAB3855MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PAR1/Thrombin Receptor Monoclonal specifically detects PAR1/Thrombin Receptor in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot.

Specifications

Antigen PAR1/Thrombin Receptor
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 731115
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CF2R, CF2RHTR, coagulation factor II (thrombin) receptor, Coagulation factor II receptor, HTR, PAR1, PAR1PAR-1protease-activated receptor 1, Protease-Activated Receptor 1, Thrombin receptor, TRproteinase-activated receptor 1
Host Species Mouse
Immunogen E. coli-derived recombinant human PAR1, Arg27-Thr102, Ser375-Thr425, Accession # P25116
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2149
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.