Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAR2 Antibody (344222), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB371144 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371144 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371144 Supplier Novus Biologicals Supplier No. FAB3949D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PAR2 Monoclonal specifically detects PAR2 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen PAR2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 344222
Conjugate DyLight 350
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human PAR2, Accession # P55085
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2150
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.