Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAR2 Antibody, Novus Biologicals™
SDP

Catalog No. NB018833 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018833 0.025 mg
NBP21125005 0.05 mg
NBP211250 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018833 Supplier Novus Biologicals Supplier No. NBP2112500.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAR2 Polyclonal specifically detects PAR2 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen PAR2
Applications Western Blot, Immunoprecipitation, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:250, ELISA 1:1500-1:20000, Immunohistochemistry 1:100, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Alias coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1
Gene Symbols F2RL1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 300-370 of mouse PAR2.
Molecular Weight of Antigen 50 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2150
Test Specificity This antibody detects a band of approximately 50kDa PAR2 protein in samples and a 52kDa protein in rat hippocampal samples. The antibody also labels a faint band of 110kDa in hippocampal membranes, probably represents either an uncharacterized variant of PAR2 or a non-specific protein.
Target Species Mouse, Rat, Human, Monkey
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.