Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS PROMO

Catalog No. J61899APF25
Click to view available options
:
500 mL

Paraformaldehyde, CAS Number-30525-89-4, methylene oxide, paraformaldehyde, formalin, paraform, methyl aldehyde, formic aldehyde, formol, oxymethylene, methanal, oxomethane.

  • Applications: Paraformaldehyde, 4% in PBS is used to preserve tissues for routine histology.
  • Solubility: Miscible with water.
  • Notes: Store in a cool place. Incompatible with brass, steel, copper, acid anhydrides, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026 g/mol
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Liquid
Quantity 500 mL
Solubility Information Miscible with Water.
Chemical Name or Material Paraformaldehyde
Hazard Category Serious Eye Damage/Eye Irritation (Category 1), Carcinogenicity (Category 2), Skin Sensitisation (Category 1)
Hazard Statement Causes Serious Eye Damage, Suspected of Causing Cancer, May Cause an Allergic Skin Reaction
TSCA No
Recommended Storage Keep Cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.