Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parainfluenza Virus type 2 Antibody (1241), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB207203 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB207203 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB207203 Supplier Novus Biologicals Supplier No. NB600834V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Parainfluenza Virus type 2 Monoclonal antibody specifically detects Parainfluenza Virus type 2 in Virus samples. It is validated for Immunofluorescence

Specifications

Antigen Parainfluenza Virus type 2
Applications Immunofluorescence
Classification Monoclonal
Clone 1241
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Human Parainfluenza Virus type 2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.