Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paramyxovirus SV5 Pk Antibody (SV5-Pk1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N1062264A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1062264A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1062264A32 Supplier Novus Biologicals Supplier No. NB10062264AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Paramyxovirus SV5 Pk Monoclonal specifically detects Paramyxovirus SV5 Pk in Virus samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Radioimmune Assays (RIA), Sandwich ELISA, Western Blot.

Specifications

Antigen Paramyxovirus SV5 Pk
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Radioimmune Assays (RIA), Sandwich ELISA, Western Blot
Classification Monoclonal
Clone SV5-Pk1
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Sandwich ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Simian virus 5; SPV5gp2; V protein
Host Species Mouse
Immunogen Mice were infected with the paramyxovirus SV5, Simian-Virus 5
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This product recognises a small epitope, termed Pk, present on the P/V proteins of the paramyxovirus, SV5. NB100-62264 has been used to detect recombinant proteins, some of which include transmembrane and secreted proteins, which have been tagged with this epitope. Usually, a 14 amino acid tag has been added to the recombinant proteins, although a smaller epitope of 9 amino acids (that as a peptide inhibit the binding of the monoclonal antibody to its native protein) has also been successfully used. The 14 amino acid epitope is; gly lys pro (ile pro asn pro leu leu gly leu asp) ser thr. (9 amino acid epitope is in parenthesis)
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.