Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paramyxovirus SV5 Pk Antibody (SV5-Pk1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017451 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017451 0.025 mg
NB10062264 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017451 Supplier Novus Biologicals Supplier No. NB100622640.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 23 publications

Paramyxovirus SV5 Pk Monoclonal specifically detects Paramyxovirus SV5 Pk in Mouse, Virus samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Radioimmune Assays (RIA), Sandwich ELISA, Western Blot.

Specifications

Antigen Paramyxovirus SV5 Pk
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Radioimmune Assays (RIA), Sandwich ELISA, Western Blot
Classification Monoclonal
Clone SV5-Pk1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Flow Cytometry 1:10-1:1000, ELISA 1:1000-1:5000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500RadioImmunoassay 1:100-1:2000, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunoassay 1:100-1:2000, Sandwich ELISA
Formulation PBS with 0.09% Sodium Azide
Gene Alias Simian virus 5; SPV5gp2; V protein
Host Species Mouse
Immunogen Mice were infected with the paramyxovirus SV5, Simian-Virus 5
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Epitope Tags, V5 Epitope Tags
Primary or Secondary Primary
Test Specificity This product recognises a small epitope, termed Pk, present on the P/V proteins of the paramyxovirus, SV5. NB100-62264 has been used to detect recombinant proteins, some of which include transmembrane and secreted proteins, which have been tagged with this epitope. Usually, a 14 amino acid tag has been added to the recombinant proteins, although a smaller epitope of 9 amino acids (that as a peptide inhibit the binding of the monoclonal antibody to its native protein) has also been successfully used. The 14 amino acid epitope is; gly lys pro (ile pro asn pro leu leu gly leu asp) ser thr. (9 amino acid epitope is in parenthesis)
Target Species Mouse, Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.