Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Paramyxovirus SV5 Pk Antibody (SV5-Pk1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 23 publications
$206.50 - $512.00
Specifications
Antigen | Paramyxovirus SV5 Pk |
---|---|
Clone | SV5-Pk1 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:10-1:1000, ELISA 1:1000-1:5000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500RadioImmunoassay 1:100-1:2000, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunoassay 1:100-1:2000, Sandwich ELISA |
Applications | Immunohistochemistry, Immunocytochemistry, Flow Cytometry, Immunofluorescence, Immunoprecipitation, ELISA, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017451
![]() |
Novus Biologicals
NB100622640.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10062264
![]() |
Novus Biologicals
NB10062264 |
0.1 mg |
Each for $512.00
|
|
|||||
Description
Paramyxovirus SV5 Pk Monoclonal specifically detects Paramyxovirus SV5 Pk in Mouse, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Radioimmunoassay, Sandwich ELISA.Specifications
Paramyxovirus SV5 Pk | |
1.0 mg/mL | |
Immunohistochemistry, Immunocytochemistry, Flow Cytometry, Immunofluorescence, Immunoprecipitation, ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Epitope Tags, V5 Epitope Tags | |
Simian virus 5; SPV5gp2; V protein | |
IgG2a | |
Protein G purified | |
This product recognises a small epitope, termed Pk, present on the P/V proteins of the paramyxovirus, SV5. NB100-62264 has been used to detect recombinant proteins, some of which include transmembrane and secreted proteins, which have been tagged with this epitope. Usually, a 14 amino acid tag has been added to the recombinant proteins, although a smaller epitope of 9 amino acids (that as a peptide inhibit the binding of the monoclonal antibody to its native protein) has also been successfully used. The 14 amino acid epitope is; gly lys pro (ile pro asn pro leu leu gly leu asp) ser thr. (9 amino acid epitope is in parenthesis) |
SV5-Pk1 | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:10-1:1000, ELISA 1:1000-1:5000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500RadioImmunoassay 1:100-1:2000, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunoassay 1:100-1:2000, Sandwich ELISA | |
Monoclonal | |
Purified | |
RUO | |
Virus, Mouse | |
Mice were infected with the paramyxovirus SV5, Simian-Virus 5 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title