Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARD3B Antibody, Novus Biologicals™
SDP

Catalog No. NB430913 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB430913 25 μL
NBP189114 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB430913 Supplier Novus Biologicals Supplier No. NBP18911425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PARD3B Polyclonal specifically detects PARD3B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PARD3B
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias ALS2CR19, amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 19, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 19 protein, MGC16131, par-3 partitioning defective 3 homolog B (C. elegans), PAR3B, PAR3beta, PAR3-beta, PAR3L, PAR3-L protein, Par3Lb, PAR3LC, partitioning defective 3 homolog B, Partitioning defective 3-like protein, partitioning-defective 3-beta
Gene Symbols PARD3B
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:ARREGFPLYEDDEGRARPSEYDLLWVPGRGPDGNAHNLRFEGMERQYASLPRGGPADPVDYLPAAPRGLYKERELPYYPGAHPMHPPKGSYPRPTELRVADLRYPQ
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 117583
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.