Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARD3B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PARD3B |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PARD3B Polyclonal specifically detects PARD3B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PARD3B | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
ALS2CR19, amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 19, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 19 protein, MGC16131, par-3 partitioning defective 3 homolog B (C. elegans), PAR3B, PAR3beta, PAR3-beta, PAR3L, PAR3-L protein, Par3Lb, PAR3LC, partitioning defective 3 homolog B, Partitioning defective 3-like protein, partitioning-defective 3-beta | |
PARD3B | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
117583 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ARREGFPLYEDDEGRARPSEYDLLWVPGRGPDGNAHNLRFEGMERQYASLPRGGPADPVDYLPAAPRGLYKERELPYYPGAHPMHPPKGSYPRPTELRVADLRYPQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title