Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARD6A Antibody, Novus Biologicals™
SDP

Catalog No. NB300953 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300953 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300953 Supplier Novus Biologicals Supplier No. NB300953
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PARD6A Polyclonal specifically detects PARD6A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen PARD6A
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry, Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_058644.
Gene Alias PAR-6, PAR-6 alpha, par-6 partitioning defective 6 homolog alpha (C. elegans), PAR6A, PAR6alpha, PAR-6Apar-6 (partitioning defective 6, C.elegans) homolog alpha, PAR6C, partitioning defective 6 homolog alpha, partitioning defective-6 homolog alpha, partitioning-defective protein 6, Tax interaction protein 40, TAX40, Tax-interacting protein 40, TIP-40PAR6
Gene Symbols PARD6A
Host Species Goat
Immunogen Peptide with sequence C-GSRIRGDGSGFSL corresponding to C-Terminus according to NP_058644.1, NP_001032358.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 50855
Test Specificity This antibody is expected to recognize both reported isoforms (NP_058644.1; NP_001032358.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.