Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARD6A Antibody (SR1623), Novus Biologicals™
SDP

Catalog No. NB259987 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259987 50 μg
NB259375 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259987 Supplier Novus Biologicals Supplier No. NBP32191050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PARD6A Monoclonal antibody specifically detects PARD6A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen PARD6A
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone SR1623
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias PAR-6, PAR-6 alpha, par-6 partitioning defective 6 homolog alpha (C. elegans), PAR6A, PAR6alpha, PAR-6Apar-6 (partitioning defective 6, C.elegans) homolog alpha, PAR6C, partitioning defective 6 homolog alpha, partitioning defective-6 homolog alpha, partitioning-defective protein 6, Tax interaction protein 40, TAX40, Tax-interacting protein 40, TIP-40PAR6
Host Species Rabbit
Immunogen A synthesized peptide derived from human PARD6A (Uniprot #: Q9NPB6)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 50855
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.