Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARG1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP105988 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105988 Supplier Novus Biologicals Supplier No. NBP105988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PARG1 Polyclonal specifically detects PARG1 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen PARG1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 1:10-1:500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_004806.3
Gene Alias PARG1PTPL1-associated RhoGAP 1 (PARG1), PTPL1-associated RhoGAP protein 1, Rho GTPase activating protein 29, rho GTPase-activating protein 29, Rho-type GTPase-activating protein 29, RP11-255E17.1
Gene Symbols ARHGAP29
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1125 and 1175 of human PTPL1-associated RhoGAP 1 using the numbering given in entry NP_004806.3 (GeneID 9411).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9411
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.