Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Park7/DJ-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB300270 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300270 0.1 mL
NB300270SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300270 Supplier Novus Biologicals Supplier No. NB300270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 19 publications

Park7/DJ-1 Polyclonal specifically detects Park7/DJ-1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, In vitro assay.

Specifications

Antigen Park7/DJ-1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In vitro Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000, Simple Western 1:4000, Flow Cytometry reported in scientific literature (PMID 21937704), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin, In vitro assay reported in scientific literature (PMID 21645620)
Formulation Whole antisera with No Preservative
Gene Accession No. Q99497
Gene Alias DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1
Gene Symbols PARK7
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus (within residues 150-189) of human Park7(DJ-1). [UniProt# Q99497]
Molecular Weight of Antigen 25 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 11315
Test Specificity Human and mouse. Predicted to react with rat, bovine, Zebra fish, Golden Syrian hamster, and chicken based on 100% sequence homology. Chicken reactivity reported in scientific literature (PMID: 24064392).
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.