Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Park7/DJ-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 19 publications
$206.50 - $501.50
Specifications
Antigen | Park7/DJ-1 |
---|---|
Dilution | Western Blot 1:2000, Simple Western 1:4000, Flow Cytometry reported in scientific literature (PMID 21937704), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin, In vitro assay reported in scientific literature (PMID 21645620) |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In vitro Assay |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300270SS
![]() |
Novus Biologicals
NB300270SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB300270
![]() |
Novus Biologicals
NB300270 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
Park7/DJ-1 Polyclonal specifically detects Park7/DJ-1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, In vitro assay.Specifications
Park7/DJ-1 | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In vitro Assay | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors | |
Whole antisera with No Preservative | |
DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1 | |
PARK7 | |
IgG | |
Unpurified | |
Human and mouse. Predicted to react with rat, bovine, Zebra fish, Golden Syrian hamster, and chicken based on 100% sequence homology. Chicken reactivity reported in scientific literature (PMID: 24064392). |
Western Blot 1:2000, Simple Western 1:4000, Flow Cytometry reported in scientific literature (PMID 21937704), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin, In vitro assay reported in scientific literature (PMID 21645620) | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat, Chicken | |
Q99497 | |
11315 | |
A synthetic peptide corresponding to the C-terminus (within residues 150-189) of human Park7(DJ-1). [UniProt# Q99497] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
25 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title