Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Park7/DJ-1 Antibody (1E12), FITC, Novus Biologicals™
SDP

Catalog No. NB160617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160617 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160617 Supplier Novus Biologicals Supplier No. NBP259407F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Park7/DJ-1 Monoclonal antibody specifically detects Park7/DJ-1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen Park7/DJ-1
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1000000000000
Conjugate FITC
Formulation PBS
Gene Alias DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1
Host Species Mouse
Immunogen Recombinant human Park7/DJ-1 (1-189aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 11315
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.