Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Park7/DJ-1 Antibody (4H4), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB344410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344410 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344410 Supplier Novus Biologicals Supplier No. NBP192715MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Park7/DJ-1 Monoclonal antibody specifically detects Park7/DJ-1 in Human, Bovine, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, KnockDown

Specifications

Antigen Park7/DJ-1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown
Classification Monoclonal
Clone 4H4
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias DJ-1, DJ1FLJ34360, EC 3.4, FLJ27376, FLJ92274, Oncogene DJ1, Parkinson disease (autosomal recessive, early onset) 7, Parkinson disease protein 7, protein DJ-1
Host Species Mouse
Immunogen Full length recombinant human Park7(DJ-1) expressed in and purified from E. coli. [UniProt# Q99497]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, DNA replication Transcription Translation and Splicing, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 11315
Target Species Human, Bovine, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.