Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parkin Antibody (PRK8), Novus Biologicals™
SDP

Catalog No. NB019485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019485 0.025 mL
NBP229838 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019485 Supplier Novus Biologicals Supplier No. NBP2298380.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Parkin Monoclonal specifically detects Parkin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Parkin
Applications Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone PRK8
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500
Formulation Ascites, containing 30% glycerol. with 0.05% Sodium Azide
Gene Accession No. O60260
Gene Alias E3 ubiquitin ligase, E3 ubiquitin-protein ligase parkin, EC 6.3.2.-, LPRS2, parkin, parkin 2, Parkinson disease protein 2, Parkinson juvenile disease protein 2, parkinson protein 2, E3 ubiquitin protein ligase (parkin), PDJjuvenile) 2, parkin
Gene Symbols PRKN
Host Species Mouse
Immunogen Full length recombinant human Parkin. Epitope has been mapped to the second ring domain (aa 399-465).
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5071
Test Specificity Parkin 52kDa.May be observed as a doublet at 52 and 46kDa weight may be detected.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.