Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parkin Mouse anti-Human, Rat, Mouse, Clone: PRK8, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB5512I25UL
Description
Parkin Monoclonal antibody specifically detects Parkin in Human, Rat, Mouse samples. It is validated for Western Blotting, ChIP assay (ChIP).
Specifications
| Parkin | |
| Monoclonal | |
| 0.5 mg/mL | |
| Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| Full length recombinant human Parkin. | |
| 25 μL | |
| Primary | |
| Stable for 1 year at +2°C to +8°C from date of receipt. | |
| IgG2b |
| ChIP Assay | |
| PRK8 | |
| Unconjugated | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human, Rat, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction