Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parkin Mouse anti-Human, Rat, Mouse, Clone: PRK8, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $468.00
Specifications
Antigen | Parkin |
---|---|
Clone | PRK8 |
Concentration | 0.5 mg/mL |
Applications | ChIP Assay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5512I25
|
MilliporeSigma
MAB5512I25UL |
25 μL |
Each for $182.31
|
|
|||||
MAB5512I100
|
MilliporeSigma
MAB5512I100UL |
100 μL |
Each for $468.00
|
|
|||||
Description
Parkin Monoclonal antibody specifically detects Parkin in Human, Rat, Mouse samples. It is validated for Western Blotting, ChIP assay (ChIP).Specifications
Parkin | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG2b | |
Protein G purified |
PRK8 | |
ChIP Assay | |
Unconjugated | |
Mouse | |
Human, Rat, Mouse | |
Full length recombinant human Parkin. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title