Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARL Antibody, Novus Biologicals™
SDP

Catalog No. NBP144262 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144262 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144262 Supplier Novus Biologicals Supplier No. NBP144262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

PARL Polyclonal specifically detects PARL in Human, Mouse, Rat, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PARL
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.86 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:20000-1:60000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with No Preservative
Gene Accession No. Q9H300
Gene Alias EC 3.4.21.105, mitochondrial, presenilin associated, rhomboid-like, PRO2207, rhomboid 7 homolog 1
Gene Symbols PARL
Host Species Rabbit
Immunogen PARL Antibody was prepared from whole rabbit serum produced by repeated immunization from a synthetic peptide corresponding to the C terminus region of human PARL. (Uniprot: Q9H300)
Molecular Weight of Antigen 42.19 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Mitochondrial Fusion Proteins, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 55486
Test Specificity This antibody is specific to PARL
Reconstitution Reconstitute with 185 ml of water to desired concentration and for immediate usage.
Target Species Human, Mouse, Rat, Chicken, Primate
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.