Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP Antibody (194C1439) - Cleaved - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP227335SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227335SS 0.025 mg
NBP227335 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227335SS Supplier Novus Biologicals Supplier No. NBP227335SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 7 publications

PARP Monoclonal specifically detects PARP in Human, Mouse, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.

Specifications

Antigen PARP
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone 194C1439
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2ug/ml∼. Use reported in scientific literature (PMID 27470599), Flow Cytometry 1ul/1 million cells, Immunocytochemistry/Immunofluorescence 2-5 ug/ml, CyTOF-ready
Gene Accession No. P09874
Gene Alias ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase), ADPRT 1, ADPRTADP-ribosyltransferase NAD(+), EC 2.4.2, EC 2.4.2.30, NAD(+) ADP-ribosyltransferase 1, PARP apoptosis, PARP-1, PARPADPRT1, poly (ADP-ribose) polymerase 1, poly (ADP-ribose) polymerase family, member 1, poly [ADP-ribose] polymerase 1, poly(ADP-ribose) polymerase, poly(ADP-ribose) synthetase, poly(ADP-ribosyl)transferase, Poly[ADP-ribose] synthase 1, PPOLpADPRT-1
Gene Symbols PARP1
Host Species Mouse
Immunogen This antibody was developed by immunizing mice with a synthetic peptide containing amino acids near 214/215-cleavage site of human PARP.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Hypoxia, Phospho Specific, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 142
Test Specificity This monoclonal antibody specifically detects a 89-kDa cleavage fragment containing the catalytic and auto modification domain.
Target Species Human, Mouse, Rat (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.