Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP Antibody (194C1439) - Cleaved - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications
$192.47 - $529.95
Specifications
| Antigen | PARP |
|---|---|
| Clone | 194C1439 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-2ug/ml∼. Use reported in scientific literature (PMID 27470599), Flow Cytometry 1ul/1 million cells, Immunocytochemistry/Immunofluorescence 2-5 ug/ml, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227335SS
![]() |
Novus Biologicals
NBP227335SS |
0.025 mg |
Each for $192.47
|
|
|||||
NBP227335
![]() |
Novus Biologicals
NBP227335 |
0.1 mg |
Each for $529.95
|
|
|||||
Description
PARP Monoclonal specifically detects PARP in Human, Mouse, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
| PARP | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Hypoxia, Phospho Specific, Stem Cell Markers, Tumor Suppressors | |
| P09874 | |
| 142 | |
| This antibody was developed by immunizing mice with a synthetic peptide containing amino acids near 214/215-cleavage site of human PARP. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 194C1439 | |
| Western Blot 1-2ug/ml∼. Use reported in scientific literature (PMID 27470599), Flow Cytometry 1ul/1 million cells, Immunocytochemistry/Immunofluorescence 2-5 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat (Negative) | |
| ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase), ADPRT 1, ADPRTADP-ribosyltransferase NAD(+), EC 2.4.2, EC 2.4.2.30, NAD(+) ADP-ribosyltransferase 1, PARP apoptosis, PARP-1, PARPADPRT1, poly (ADP-ribose) polymerase 1, poly (ADP-ribose) polymerase family, member 1, poly [ADP-ribose] polymerase 1, poly(ADP-ribose) polymerase, poly(ADP-ribose) synthetase, poly(ADP-ribosyl)transferase, Poly[ADP-ribose] synthase 1, PPOLpADPRT-1 | |
| PARP1 | |
| IgG2b κ | |
| Protein G purified | |
| This monoclonal antibody specifically detects a 89-kDa cleavage fragment containing the catalytic and auto modification domain. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title