Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP16 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB133905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB133905 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB133905 Supplier Novus Biologicals Supplier No. NBP306231AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PARP16 Polyclonal specifically detects PARP16 in Human samples. It is validated for Western Blot.

Specifications

Antigen PARP16
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias C15orf30, chromosome 15 open reading frame 30, EC 2.4.2.30, FLJ20509, FLJ25281, PARP-16, pART15, poly (ADP-ribose) polymerase family, member 16, poly [ADP-ribose] polymerase 16
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PARP16 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 54956
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.