Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP2 Antibody (4G8), Novus Biologicals™
SDP

Catalog No. NB036632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036632 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036632 Supplier Novus Biologicals Supplier No. NBP289041
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PARP2 Monoclonal specifically detects PARP2 in Human, Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PARP2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4G8
Conjugate Unconjugated
Formulation Tissue culture supernatant
Gene Alias ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2, ADPRT-2, ADPRT2pADPRT-2, ADPRTL2EC 2.4.2.30, ADPRTL3, hPARP-2, NAD(+) ADP-ribosyltransferase 2, PARP-2, poly (ADP-ribose) polymerase 2, poly (ADP-ribose) polymerase family, member 2, poly (ADP-ribosyl) transferase-like 2, poly [ADP-ribose] polymerase 2, poly(ADP-ribose) synthetase, Poly[ADP-ribose] synthase 2, poly[ADP-ribose] synthetase 2
Host Species Mouse
Immunogen Recombinant mouse PARP2 (poly(ADP-ribose) polymerase-2).
Quantity 500 μL
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 10038
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.