Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP2 Antibody - Unpurified, Novus Biologicals™
SDP

Catalog No. NB000932 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000932 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000932 Supplier Novus Biologicals Supplier No. NB100185SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PARP2 Polyclonal specifically detects PARP2 in Human samples. It is validated for Western Blot.

Specifications

Antigen PARP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:250-1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2, ADPRT-2, ADPRT2pADPRT-2, ADPRTL2EC 2.4.2.30, ADPRTL3, hPARP-2, NAD(+) ADP-ribosyltransferase 2, PARP-2, poly (ADP-ribose) polymerase 2, poly (ADP-ribose) polymerase family, member 2, poly (ADP-ribosyl) transferase-like 2, poly [ADP-ribose] polymerase 2, poly(ADP-ribose) synthetase, Poly[ADP-ribose] synthase 2, poly[ADP-ribose] synthetase 2
Gene Symbols PARP2
Host Species Rabbit
Immunogen Residues 43-59 [QRQESKKMPVAGGKANK] of the 62 kDa human PARP-2 protein.
Molecular Weight of Antigen 66 kDa
Purification Method Whole Antiserum
Quantity 0.025 mL
Research Discipline Apoptosis, DNA Repair, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 10038
Test Specificity N-terminus of human PARP 2
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.