Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 112891AP150UL

Description
PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintece of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.Specifications
PARP3 | |
Polyclonal | |
Unconjugated | |
PARP3 | |
ADPRT 3, ADPRT3, ADPRTL2, ADPRTL3, hPARP 3, IRT1, PADPRT 3, PARP 3, PARP3, Poly [ADP ribose] polymerase 3, Poly[ADP ribose] synthase 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10039, 235587, 300985 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3ULW8, Q9Y6F1 | |
PARP3 | |
PARP3 Fusion Protein Ag1844 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction