Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PARP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PARP3 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7034
![]() ![]() |
Proteintech Group Inc
112891AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
PARP3, poly(ADP-ribosyl)transferase 3, belongs to the PARP family. These enzymes modify nuclear proteins by poly-ADP-ribosylation, which is required for DNA repair, regulation of apoptosis, and maintece of genomic stability. This protein is preferentially localized to the daughter centriole throughout the cell cycle.Specifications
PARP3 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q3ULW8, Q9Y6F1 | |
10039, 235587, 300985 | |
PARP3 Fusion Protein Ag1844 | |
Primary | |
-20°C | |
PARP3 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ADPRT 3, ADPRT3, ADPRTL2, ADPRTL3, hPARP 3, IRT1, PADPRT 3, PARP 3, PARP3, Poly [ADP ribose] polymerase 3, Poly[ADP ribose] synthase 3 | |
PARP3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title