Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP4 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB046931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB046931 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB046931 Supplier Novus Biologicals Supplier No. NBP297710R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PARP4 Polyclonal specifically detects PARP4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PARP4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 193 kDa vault protein, ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 1, ADPRTL1I-alpha-I-related, EC 2.4.2.30, H5 proline-rich, p193, PARP-4, PARPL, PARP-related, PARP-related/IalphaI-related H5/proline-rich, PH5P, poly (ADP-ribose) polymerase family, member 4, poly [ADP-ribose] polymerase 4, poly(ADP-ribose) synthetase, poly(ADP-ribosyl)transferase-like 1, Vault poly(ADP-ribose) polymerase, vault protein, 193-kDa, VAULT3, von Willebrand factor A domain containing 5C, VPARPKIAA0177, VWA5C
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PARP4 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 143
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.