Learn More
Description
- Straight-sided container format featuring top and bottom positive grip knurls for ease of opening and closing when wearing protective or latex gloves
- Stackable, leak-resistant and graduated unless otherwise noted
- Sterile containers labeled with a security tab label and sterilized by gamma irradiation to SAL 10-3; certification of sterility available upon request
- Manufactured from virgin polypropylene (container) and virgin polyethylene (closure) without the use of plasticizers or demolding additives; all material used is free from latex and bovine contamination
- Manufactured under strict process control and lot numbers are fully traceable
- 95kpa compliant; testing results available upon request
- Rigorous testing at varying pressures without seal failure proves this container ideal for use in pneumatic tube systems and harsh transport conditions; leak performance data is available upon request
- Available in individually wrapped (Flo-Pac™) formats
- Closures secure at 10 to 12 in.-lb. closing force
- Reagent or fixative pre-fills may require 15 to 17 in.-lb. closing force to prevent post-production cap-back-off
- Standard cap colors are natural, orange and blue
Specifications
Specifications
| Graduated | Yes |
| Material | Polypropylene (Clarified) |
| Sterility | Sterile |
| Capacity (English) | 4 oz. |
| Capacity (Metric) | 120 mL |
| Closure Color | Blue |
| Closure Material | Polyethylene |
| Closure Size | 53 mm |
| Label | Tamper-evident Label |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.