Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parvalbumin Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB338938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338938 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338938 Supplier Novus Biologicals Supplier No. NB12011427APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Parvalbumin Polyclonal antibody specifically detects Parvalbumin in Human, Mouse, Rat, Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Free-Floating

Specifications

Antigen Parvalbumin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating
Formulation PBS
Gene Alias D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV
Host Species Rabbit
Immunogen Parvalbumin Antibody made from purified parvalbumin
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5816
Target Species Human, Mouse, Rat, Feline
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.