Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parvalbumin Antibody (C12), Novus Biologicals™
SDP

Catalog No. NB010674 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010674 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010674 Supplier Novus Biologicals Supplier No. NBP279824
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Parvalbumin Monoclonal specifically detects Parvalbumin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Parvalbumin
Applications Western Blot, ELISA
Classification Monoclonal
Clone C12
Conjugate Unconjugated
Dilution Western Blot, ELISA
Formulation PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Gene Alias D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV
Gene Symbols PVALB
Host Species Mouse
Immunogen Synthetic peptide from the full length Rat Parvalbumin protein
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20 °C.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.