Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARVB Antibody, Novus Biologicals™
SDP

Catalog No. NBP15913520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15913520 20 μL
NBP159135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15913520 Supplier Novus Biologicals Supplier No. NBP15913520UL

Rabbit Polyclonal Antibody

PARVB Polyclonal specifically detects PARVB in Human samples. It is validated for Western Blot.

Specifications

Antigen PARVB
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96PN1
Gene Alias affixin, beta-parvin, CGI-56, parvin, beta
Gene Symbols PARVB
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PARVB(parvin, beta) The peptide sequence was selected from the C terminal of PARVB. Peptide sequence HNVSFAFELMLDGGLKKPKARPEDVVNLDLKSTLRVLYNLFTKYKNVE.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 29780
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.