Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parvin gamma Antibody (8C5.2), Biotin, Novus Biologicals™
SDP

Catalog No. NB391119 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391119 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391119 Supplier Novus Biologicals Supplier No. NBP308728B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Parvin gamma Monoclonal antibody specifically detects Parvin gamma in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Parvin gamma
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 8C5.2
Conjugate Biotin
Formulation PBS
Gene Alias gamma-parvin, parvin, gamma
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Parvin gamma (Uniprot: Q9HBI0)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64098
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.