Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parvovirus NS1 Antibody (8E6E10) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP261903

 View more versions of this product

Catalog No. NBP2-61903


Only null left
Add to Cart

Description

Description

Parvovirus NS1 Monoclonal specifically detects Parvovirus NS1 in Human, Viral samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Parvovirus NS1
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
NS1
Purified recombinant fragment of human Parvovirus NS1 (AA: 1-216) expressed in E. Coli.
Protein G purified
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
8E6E10
Unconjugated
PBS with 0.05% Sodium Azide
Mouse
585 kDa
0.1 mL
Human, Virus
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.