Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Parvovirus NS1 Antibody (8E6E10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Clone | 8E6E10 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61903
![]() |
Novus Biologicals
NBP261903 |
0.1 mL |
Each of 1 for $560.50
|
|
|||||
Description
Parvovirus NS1 Monoclonal specifically detects Parvovirus NS1 in Human, Viral samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
8E6E10 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
Monoclonal | |
Purified | |
Human, Virus | |
Purified recombinant fragment of human Parvovirus NS1 (AA: 1-216) expressed in E. Coli. | |
Primary | |
585 kDa |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
NS1 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title