Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PASD1 Antibody (2ALCC128), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP250234FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250234FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250234FR Supplier Novus Biologicals Supplier No. NBP250234FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PASD1 Monoclonal specifically detects PASD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen PASD1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Monoclonal
Clone 2ALCC128
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CT63Cancer/testis antigen 63, OXTES1, OX-TES-1, PAS domain containing 1, PAS domain-containing protein 1
Gene Symbols PASD1
Host Species Mouse
Immunogen GST-PASD1 recombinant protein containing the 540-773 aas of PASD1, a region only present in PASD1b.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 139135
Test Specificity Specific for the recombinant protein (aa 540-773) present only in the longer PASD1b protein (C-terminus)
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.