Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Patched 1/PTCH Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP171662A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP171662A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP171662A70 Supplier Novus Biologicals Supplier No. NBP171662AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Patched 1/PTCH Polyclonal specifically detects Patched 1/PTCH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Patched 1/PTCH
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BCNSFLJ42602, FLJ26746, HPE7, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4′, PTCH protein -10, PTCH11
Gene Symbols PTCH1
Host Species Rabbit
Immunogen A genomic peptide made to a C-terminal region of the human Patched 1 protein (within residues 1200-1350). [Swiss-Prot Q13635]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5727.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.