Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Patched 1/PTCH Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP171662C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171662C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP171662C Supplier Novus Biologicals Supplier No. NBP171662C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Patched 1/PTCH Polyclonal specifically detects Patched 1/PTCH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Patched 1/PTCH
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q13635
Gene Alias BCNSFLJ42602, FLJ26746, HPE7, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4', PTCH protein -10, PTCH11
Gene Symbols PTCH1
Host Species Rabbit
Immunogen A genomic peptide made to a C-terminal region of the human Patched 1 protein (within residues 1200-1350). [Swiss-Prot Q13635]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5727
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.