Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Patched 1/PTCH Antibody, Novus Biologicals™
SDP

Catalog No. NB10041101 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10041101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10041101 Supplier Novus Biologicals Supplier No. NB10041101
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Patched 1/PTCH Polyclonal specifically detects Patched 1/PTCH in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA, Western Blot.

Specifications

Antigen Patched 1/PTCH
Applications Immunocytochemistry, Immunofluorescence, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunocytochemistry/Immunofluorescence, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000255.1
Gene Alias BCNSFLJ42602, FLJ26746, HPE7, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4', PTCH protein -10, PTCH11
Gene Symbols PTCH1
Host Species Goat
Immunogen Peptide with sequence C-HPESRHHPPSNPRQQ corresponding to internal region according to NP_000255.2, NP_001077072.1, NP_001077071.1, NP_001077075.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5727
Test Specificity This antibody is expected to recognize all four reported isoforms (NP_000255.2; NP_001077072.1; NP_001077071.1; NP_001077075.1). Reported variants represent identical protein (NP_001077075.1; NP_001077076.1; NP_001077073.1; NP_001077074.1).
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.