Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Patched 1/PTCH Antibody (OTI1H10), PE, Novus Biologicals™
SDP

Catalog No. NB302079 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302079 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302079 Supplier Novus Biologicals Supplier No. NBP273254PE

Mouse Monoclonal Antibody

Patched 1/PTCH Monoclonal antibody specifically detects Patched 1/PTCH in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Patched 1/PTCH
Applications Western Blot
Classification Monoclonal
Clone OTI1H10
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias BCNSFLJ42602, FLJ26746, HPE7, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4', PTCH protein -10, PTCH11
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 770-1027 of human PTCH1 (NP_000255) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5727
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.