Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Patched 2/PTCH2 Antibody (OTI8B5), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB302078 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302078 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302078 Supplier Novus Biologicals Supplier No. NBP273255APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Patched 2/PTCH2 Monoclonal antibody specifically detects Patched 2/PTCH2 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Patched 2/PTCH2
Applications Western Blot
Classification Monoclonal
Clone OTI8B5
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias patched (Drosophila) homolog 2, patched 2, patched homolog 2, patched homolog 2 (Drosophila), protein patched homolog 2, PTC2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 79-392 of human PTCH2 (NP_003729) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8643
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.