Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Patched 2/PTCH2 Antibody (OTI8B5), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP273255UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273255UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273255UV Supplier Novus Biologicals Supplier No. NBP273255UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Patched 2/PTCH2 Monoclonal specifically detects Patched 2/PTCH2 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Patched 2/PTCH2
Applications Western Blot
Classification Monoclonal
Clone OTI8B5
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias patched (Drosophila) homolog 2, patched 2, patched homolog 2, patched homolog 2 (Drosophila), protein patched homolog 2, PTC2
Gene Symbols PTCH2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 79-392 of human Patched 2/PTCH2 (NP_003729) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8643.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.