Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Patched Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB360778 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360778 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360778 Supplier Novus Biologicals Supplier No. NBP223623APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Patched Polyclonal antibody specifically detects Patched in Mouse samples. It is validated for Western Blot

Specifications

Antigen Patched
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias PTCH
Host Species Rabbit
Immunogen Reacts with residues 269-279 [KKINYQVDSWE] of the mouse PTCH protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.