Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Patched Domain Containing 2 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB164791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164791 Supplier Novus Biologicals Supplier No. NBP281757B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Patched Domain Containing 2 Polyclonal antibody specifically detects Patched Domain Containing 2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Patched Domain Containing 2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias DISP3, Dispatched 3, KIAA1337, Patched Domain-Containing Protein 2, Protein Dispatched Homolog 3, PTCHD2
Host Species Rabbit
Immunogen Patched Domain Containing 2 antibody was raised against a 19 amino acid synthetic peptide from near the amino terminus of human Patched Domain Containing 2. The immunogen is located within amino acids 210 - 260 of Patched Domain Containing 2. Amino Acid Squence: SEDPRNQRLSKNGRYQPSI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57540
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.