Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PATZ Antibody (OTI8D6), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP273256C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273256C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273256C Supplier Novus Biologicals Supplier No. NBP273256C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PATZ Monoclonal specifically detects PATZ in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PATZ
Applications Western Blot
Classification Monoclonal
Clone OTI8D6
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias dJ400N23, MAZRPOZ-, AT hook-, and zinc finger-containing protein 1, PATZZinc finger and BTB domain-containing protein 19, POZ (BTB) and AT hook containing zinc finger 1, Protein kinase A RI subunit alpha-associated protein, RIAZBTB/POZ domain zinc finger transcription factor, ZBTB19BTB-POZ domain zinc finger transcription factor, Zinc finger protein 278Zinc finger sarcoma gene protein, ZNF278POZ-AT hook-zinc finger protein, ZSGMAZ-related factor
Gene Symbols PATZ1
Host Species Mouse
Immunogen Full length recombinant protein of human PATZ (NP_114440) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23598.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.