Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAUF/ZG16B Antibody (6E5), Novus Biologicals™
SDP

Catalog No. NB298489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298489 100 μL
NB298488 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298489 Supplier Novus Biologicals Supplier No. NBP326244100UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

PAUF/ZG16B Monoclonal antibody specifically detects PAUF/ZG16B in Human samples. It is validated for ELISA

Specifications

Antigen PAUF/ZG16B
Applications ELISA
Classification Monoclonal
Clone 6E5
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias DKFZp762P2216, EC 3.1.2, EC 3.4.19.12, hTrabid, TRABIDubiquitin thioesterase ZRANB1, TRAF-binding protein domain, Zinc finger Ran-binding domain-containing protein 1, zinc finger, RAN-binding domain containing 1
Host Species Human
Immunogen Recombinant Human PAUF/ZG16B protein [UniProt Q96DA0]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124220
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.