Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAUF/ZG16B Antibody (6E5), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | PAUF/ZG16B |
---|---|
Clone | 6E5 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
PAUF/ZG16B Monoclonal antibody specifically detects PAUF/ZG16B in Human samples. It is validated for ELISASpecifications
PAUF/ZG16B | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
124220 | |
IgG1 | |
Affinity purified |
6E5 | |
ELISA | |
Unconjugated | |
Human | |
Human | |
DKFZp762P2216, EC 3.1.2, EC 3.4.19.12, hTrabid, TRABIDubiquitin thioesterase ZRANB1, TRAF-binding protein domain, Zinc finger Ran-binding domain-containing protein 1, zinc finger, RAN-binding domain containing 1 | |
Recombinant Human PAUF/ZG16B protein [UniProt Q96DA0] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title