Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAWR Antibody (SR1674), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32220850UL
Description
PAWR Monoclonal antibody specifically detects PAWR in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PAWR | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
A synthesized peptide derived from human PAWR (Uniprot #: Q96IZ0) | |
50 μg | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot | |
SR1674 | |
Western Blot 1:500-1:2000 | |
Rabbit | |
Affinity purified | |
RUO | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction