Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAWR Antibody (SR1674), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PAWR |
---|---|
Clone | SR1674 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
PAWR Monoclonal antibody specifically detects PAWR in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PAWR | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
IgG | |
Affinity purified |
SR1674 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
A synthesized peptide derived from human PAWR (Uniprot #: Q96IZ0) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title